Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Taurodeoxycholic acid, sodium salt hydrate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
207737-97-1,207737-97-1
C26H46NNaO7S
539.704
MFCD00149238
OLPIZAYYAVQETM-GGPRKOIFSA-M
Sodium taurodeoxylate
23702150
sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate;hydrate
CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C.O.[Na+]
207737-97-1,207737-97-1
C26H46NNaO7S
539.704
MFCD00149238
OLPIZAYYAVQETM-GGPRKOIFSA-M
Sodium taurodeoxylate
23702150
sodium;2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]ethanesulfonate;hydrate
CC(CCC(=O)NCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2CCC4C3(CCC(C4)O)C)O)C.O.[Na+]
Appearance of Solution (5% aq. soln.) clear colorless to faint yellow
Chloride 0.5% max.
Heavy Metals (as Pb) 20ppm max.
Assay Percent Range 97%
Infrared Spectrum Authentic
Packaging Glass bottle
Formula Weight 521.69
Physical Form Crystalline Powder
Percent Purity ≥96.0%, 1.0% max. taurine, 1.0% max. sodium taurocholate, 1.0% max. sodium deocycholate (TLC)
Specific Rotation +34.5° to +36.5° (20°C, 589nm) (c=2.5, water) (calculated on dry substance)
Water 7% Max. (K.F.)
Specific Rotation Condition +33.00 (20.00°C c=3,water)
Color White
Quantity 25g
Chemical Name or Material Taurodeoxycholic acid, sodium salt hydrate, 97%
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. Wear protective gloves/protective clothing/eye protection/face protection.

EINECSNumber : 214-652-0