Learn More
Novus Biologicals™ TAX1BP3 Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Manufacturer:  Novus Biologicals™ NBP255345PEP
Catalog No. NBP55345PEP
Add to cart
Description
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human TAX1BP3. The TAX1BP3 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications
Specifications
Antibody Competition
30851
>80% by SDS-PAGE and Coomassie blue staining
E. coli
RUO
TAX1BP3
PBS and 1M Urea, pH 7.4.
24 kDa
100 μl
Store at -20°C. Avoid freeze-thaw cycles.
TAX1BP3
Human