Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Taxol Inhibitor

Low molecular weight, organic compound that may help regulate biological process.

Specifications

For Use With (Application) Inhibits apoptosis
Solubility Soluble to 100mM in DMSO, 25mM in ethanol
Storage Requirements Store at 4°C
Target Proteins Taxol
Color White
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB109710MG View Documents Novus Biologicals™
NB109710MG
10mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
NB109750MG View Documents Novus Biologicals™
NB109750MG
50mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Biological Activity: Antitumor agent; promotes and stabilizes tubulin polymerization, causing cell cycle arrest. Induces autocatalytic activation of caspase-10 in CCRF-HSB-2 cells, triggering apoptosis.

Storage: Store at 4°C.

Note: Inhibitors are guaranteed for 1 year from date of receipt.

Specifications

Specifications

Inhibits apoptosis
Store at 4°C
White
Soluble to 100mM in DMSO, 25mM in ethanol
Taxol

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit