Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tazarotene, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Receptor-selective acetylenic retinoid; binds at retinoid acid receptors (RAR) -β and -γ. Topically used for the treatment of skin disorders, particularly psoriasis and acne. Implicated in the treatment of basal cell carcinomas; induces upregulation of Bax in vivo. Exhibits antiproliferative and proapoptotic activity. Upregulates TIGs (Tazarotene-induced genes), including the cell adhesion molecule TIG1 and the tumor-suppressor TIG3.
Specifications
Quantity | 50 mg |
Formula Weight | 351.46 |
Percent Purity | >99% |
Chemical Name or Material | Tazarotene |
Safety and Handling
Recommended Storage : Store at +4°C
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title