Learn More
Fisherbrand™ Flexible Kynar Lined Tubing
Chemical resistant Kynar lined bioprocess and medical grade tubing. More flexible than tubing extruded from 100% Kynar. Sterile using E-beam. Shore A 85
 View more versions of this product
Catalog No. 01-000-797
Add to cart
Description
Description
  • PVC-Free - No DEHP or BPA additive
  • Low extractables / leachables liner component
  • Kynar liner meets USP Class VI requirements
  • Non-Animal Derived Kynar liner
  • RoHS Compliant
  • Lot Traceable
Warranty and Services
Replacement if not satisfied. 12 Month return Policy.
Specifications
Specifications
Complaince Certificates Available
Class 7 Cleanroom Manufactured
KFlex - Kynar™ Lined Thermoplastic Polymer
Tubing
50 ft. Roll - Bag
Out of direct sunlight, >20°F <100°F
White Opaque
1/2 in.
3/4 in.
Medical, Pharmaceutical Uses
KFlex - Kynar Lined Tubing