Learn More
Cell Biolabs IncSupplier Diversity Partner TBARS ASSAY KIT TRIAL SIZE  DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA330T
The OxiSelect TBARS Assay Kit (Trial Size) offers a simple, reproducible system for the detection of lipid peroxidation in urine, plasma, serum, lysates, and tissue homogenates in a convenient 96-well format. 20 assays/kit.
Catalog No. 50-673-525