Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D16 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | TBC1D16 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP155489
|
Novus Biologicals
NBP155489 |
100 μL |
Each of 1 for $436.00
|
|
Description
TBC1D16 Polyclonal specifically detects TBC1D16 in Human samples. It is validated for Western Blot.Specifications
TBC1D16 | |
Polyclonal | |
Rabbit | |
Q8TBP0 | |
125058 | |
Synthetic peptides corresponding to TBC1D16(TBC1 domain family, member 16) The peptide sequence was selected from the middle region of TBC1D16. Peptide sequence RGEVWPFLLRYYSHESTSEEREALRLQKRKEYSEIQQKRLSMTPEEHRAF. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
RUO | |
FLJ20748, MGC25062, TBC1 domain family member 16, TBC1 domain family, member 16 | |
TBC1D16 | |
IgG | |
Affinity Purified | |
86 kDa |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title