Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D20 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549061
Description
GTPase-activating protein specific for Rab1 and Rab2 small GTPase families for which it can accelerate the intrinsic GTP hydrolysis rate by more than five orders of magnitude.Specifications
TBC1D20 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96BZ9 | |
TBC1D20 | |
KLH conjugated synthetic peptide between 335-363 amino acids from the C-terminal region of human TBC1D20. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
TBC1D20 | |
TBC1 domain family member 20, TBC1D20, C20orf140 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128637 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction