Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBC1D22A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550801
Description
The antibody detects endogenous levels of total TBC1D22A protein.
TBC1D22A Polyclonal antibody specifically detects TBC1D22A in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
TBC1D22A | |
Polyclonal | |
Unconjugated | |
TBC1D22A | |
BC023106; C22orf4; D15Ertd781e; HSC79E021; putative GTPase activator; RGD1306588; TBC1 domain family member 22A; TBC1 domain family, member 22a; TBC1D22A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25771 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1.4 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q8WUA7 | |
TBC1D22A | |
Fusion protein of human TBC1 domain family, member 22A | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction