Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ TBC1D22B Human Recombinant Protein
Highly purified. Generating reliable and reproducible results.
Supplier: Novus Biologicals™ NBP2521030.05MG
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-505 of Human TBC1D22B The Recombinant Human TBC1D22B Protein is derived from E. coli. The Recombinant Human TBC1D22B Protein has been validated for the following applications: SDS-Page.Specifications
0.25mg/mL | |
Phosphate buffered saline (pH7.4) containing 30% glycerol, 1mM DTT | |
TMW: 61.5kDa | |
0.05mg | |
C6orf197, chromosome 6 open reading frame 197, dJ744I24.2, DKFZp762J0110, FLJ20337, MGC125626, MGC125627, RP4-744I24.2, TBC1 domain family member 22B, TBC1 domain family, member 22B | |
Unlabeled |
SDS-Page | |
55633 | |
>80%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain | |
Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. | |
TBC1D22B | |
Yes |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For research use only and is not approved for use in humans or in clinical diagnosis