Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBC1D5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA557166
Description
Immunogen sequence: DVKRTFPEMQ FFQQENVRKI LTDVLFCYAR ENEQLLYKQG MHELLAPIVF VLHCDHQAFL HASESAQPSE EMKTVLNPEY LEHDAYAVFS QL Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.
TBC1 domain family member 5 (TBC1D5) may act as a GTPase-activating protein for Rab family protein(s).
Specifications
TBC1D5 | |
Polyclonal | |
Unconjugated | |
TBC1D5 | |
1600014N05Rik; KIAA0210; RGD1561626; TBC1 domain family member 5; TBC1 domain family, member 5; TBC1D5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
501088, 72238, 9779 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q80XQ2, Q92609 | |
TBC1D5 | |
Recombinant protein corresponding to Human TBC1D5. Recombinant protein control fragment (Product #RP-96332). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction