Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCA Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551186

Catalog No. PA551186


Add to cart

Description

Description

The antibody detects endogenous levels of total TBCA protein.

The product of this gene is one of four proteins (cofactors A, D, E, and C) involved in the pathway leading to correctly folded beta-tubulin from folding intermediates. Cofactors A and D are believed to play a role in capturing and stabilizing beta-tubulin intermediates in a quasi-native confirmation. Cofactor E binds to the cofactor D/beta-tubulin complex; interaction with cofactor C then causes the release of beta-tubulin polypeptides that are committed to the native state.
Specifications

Specifications

TBCA
Polyclonal
Unconjugated
TBCA
Liquid
TBCA
Full length fusion protein of human TBCA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
0.7 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
O75347, P48428
CFA, chaperonin cofactor A, TBCA, TCP1-chaperonin cofactor A, tubulin folding cofactor A, Tubulin-folding cofactor A, Tubulin-specific chaperone A
Rabbit
IgG
100 μL
-20°C
Primary
21371, 6902
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit