Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBCEL Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548374

Catalog No. PA548374

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse and rat based on sequence homology.

Acts as a regulator of tubulin stability.
Specifications

Specifications

TBCEL
Polyclonal
PBS with 0.09% sodium azide
Q5QJ74
Tubulin-specific chaperone cofactor E-like protein, EL, Leucine-rich repeat-containing protein 35, TBCEL, LRRC35
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
219899
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TBCEL
Liquid
TBCEL
KLH conjugated synthetic peptide between 385-413 amino acids from the C-terminal region of human TBCEL.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit