Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP30389220UL
Description
TBK1 Polyclonal antibody specifically detects TBK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
TBK1 | |
Unconjugated | |
Purified | |
Rabbit | |
IgG | |
20 μL | |
Phospho Specific, Protein Kinase, Signal Transduction | |
Primary | |
29110 |
Immunofluorescence, Western Blot | |
PBS with 50% glycerol, pH7.3. | |
EC 2.7.11, EC 2.7.11.1, FLJ11330, NAK serine/threonine-protein kinase TBK 1, NAKserine/threonine-protein kinase TBK1, NF-kappa-B-activating kinase, T2K, TANK-binding kinase 1NF-kB-activating kinase, TBK 1 | |
A synthetic peptide corresponding to a sequence within amino acids 100-200 of mouse TBK1 (NP_062760.3). | |
Affinity purified | |
RUO | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Polyclonal | |
Human, Mouse, Rat |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title