Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565664
Description
Immunogen sequence: RIRKEKPQQHN FTHRLLAAAL KSHSGNISCM DFSSNGKYLA TCADDRTIRI WSTKDFLQRE HRSMRANVEL DHATLVRFSP DCRAFIVWLA NGDTLRVFKM TKRE Highest antigen sequence identity to the following orthologs - mouse 96%, rat 98%.
This gene encodes a member of the beta-transducin protein family. Most proteins of the beta-transducin family are involved in regulatory functions. This protein is possibly involved in some intracellular signaling pathway. This gene is deleted in Williams-Beuren syndrome, a developmental disorder caused by deletion of multiple genes at 7q11.23.
Specifications
TBL2 | |
Polyclonal | |
Unconjugated | |
TBL2 | |
C76179; DKFZP43N024; MGC134739; TBL2; transducin (beta)-like 2; transducin beta like 2; transducin beta-like protein 2; transducin -like 2; UNQ563/PRO1125; WBSCR13; Williams-Beuren syndrome chromosomal region 13 protein; Williams-Beuren syndrome chromosome region 13; WS beta-transducin repeats protein; WS-betaTRP; WS-bTRP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26608 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9Y4P3 | |
TBL2 | |
Recombinant Human TBL2. Recombinant protein control fragment (Product #RP-106338). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction