Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBPL2 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18026820UL

 View more versions of this product

Catalog No. NBP18026820


Add to cart

Description

Description

TBPL2 Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

TBPL2
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
TBPL2
Synthetic peptide directed towards the N terminal of mouse TRF3. Peptide sequence FHPHLGGVKKASTDFSSVDLSFLPDELTQENRDQTVTGNKLASEESCRTR.
Protein A purified
RUO
Primary
387332
Western Blot
Western Blot 1:1000
NP_951014
TATA box-binding protein-like protein 2, TATA box binding protein like 2, TBP2, TRF3
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit