General Data Healthcare™ CUT™ 4055 Rotary Microtomes
Manufacturer: General Data Healthcare™ CUT4055
 View all options for this product
Catalog No. 22-046-801
  Check Availability
Add to Cart
    Safety Features Finger guards and a debris protection system
    Includes Microtome, knife holder base unit, disposable blade holder, low profile clamping plate, quick release cassette clamp, X-Y object orientation unit, plastic waste tray, a pack of low profile coated disposable blade and a 100mL bottle of PARA/GARD
    • Ergonomic design and advanced safety features are ideal for the histology lab
    • Each precision-engineered microtome is mounted on a one-piece metal base enclosed in a polycarbonate cover
    • Sturdy, dependable, low maintenance microtomes are designed to be configured for optimum user comfort with state-of-the-art accessories
    • Dual movements provide precise and efficient horizontal advance and specimen return
    • Rotary Microtomes can be ordered with the coarse advance hand wheel in the most convenient operating position according to laboratory needs
    • Positioning for the coarse advance hand wheel include side- or front-mounted with either forward or reverse feed
    • A sliding clutch over-extension protection feature automatically blocks the continued horizontal movement of the specimen after it has reached the full extent of its travel in either direction
    • The safety features of each microtome model include finger guards and a debris-protection system that protects the guideways from contamination
    • The fine advance hand wheel features a push button safety lock that can be set in 30 click positions over 360°
    • Counter-balanced for smooth sectioning, handwheel can be operated in either clockwise (CW) or counter-clockwise (CCW) feed directions
    • Auto trim can be set in three positions: 10, 20 or 30µm for quick, consistent and controlled sectioning
    • Microtomes can be purchased in a package or separately
    • Choose forward or reverse feed; side- or front-mounted handwheel
    • Sliding clutch overextension protection automatically blocks continued horizontal movement of specimen beyond the full extent of its travel in either direction
    • Fine-advance handwheel with push-button safety lock can be set in 12° increments from 0° to 360°
    • Models 4060 and 4060E feature an auto trim coarse advance hand wheel
  • Description & Specifications
  • Accessories
Related Products
Product Content Feedback