Learn More
General Data Healthcare™ PARA/Gard™ Liquid Paraffin Repellent
Pleasant smelling, non-toxic spray that keeps surfaces looking brand new
Manufacturer:  General Data Healthcare™ PG16
Catalog No. 22-047-091
Add to cart
Description
Description
  • Formulated to keep metal, wood and plastic surfaces clean
  • Used as preventive to keep paraffin from sticking to laboratory surfaces
  • Spray onto Kimwipe and then apply thin coating to effected surfaces
  • Not for use on microtome blades, blade holders or base molds
  • Available in 4 or 16 oz. (113.4 or 453g) bottles
Specifications
Specifications
Liquid Paraffin Repellent
Countertops and equipment