Learn More
General Data Healthcare™ SHUR/Mark<™ Marking Pens
SDP
Laboratory marking pens with priming action and replaceable felt tips
Supplier:  General Data Healthcare™ MPB
 View more versions of this product
Catalog No. 15-183-10
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
This item is not returnable. View return policy
Description
Description
  • For cassettes and slides
  • Fast drying
  • Simply depress tip until saturated
  • Will not smudge or smear
  • Reagent resistant to histology solvents, especially alcohol and xylene
  • Replaceable tip eliminates clogged or worn out tips
Specifications
Specifications
Marking Pen
Black
For labeling slides or cassettes
12 Pk.