Learn More
General Data Healthcare™ SHUR/Mark<™ Marking Pens
SDP
Laboratory marking pens with priming action and replaceable felt tips
Specifications
For Use With (Application) For labeling slides or cassettes
View More Specs
Products 2
Catalog Number Mfr. No. Type Price Quantity & Availability  
Catalog Number Mfr. No. Type Price Quantity & Availability  
15-183-10
SDP
View Documents
General Data Healthcare™
MPB
Marking Pen This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
22-048-205
SDP
View Documents
General Data Healthcare™
MPR
Marking Pen This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • For cassettes and slides
  • Fast drying
  • Simply depress tip until saturated
  • Will not smudge or smear
  • Reagent resistant to histology solvents, especially alcohol and xylene
  • Replaceable tip eliminates clogged or worn out tips
Specifications
Specifications
For labeling slides or cassettes