Learn More
General Data Healthcare™ SHUR/Mark™ Colormark™ Microscope Slides
Prepackaged slides in thumb-tab box for convenient handling
Manufacturer:  General Data Healthcare™ SMSCTTY
 View more versions of this product
Catalog No. 18-186-20
Add to cart
Description
Description
  • Thumb-tab cutouts in box facilitate slide pickup
  • Insert slides directly into hoppers with one easy step
  • Contrasting dark color on bottom of slides enhances writing visibility
  • Available with or without Silane coating
Specifications
Specifications
75mm
1mm
Silane
25mm
Yellow
72/Pk.