General Data Healthcare™ SHUR/Mark™ Colormark™ Microscope Slides

Prepackaged slides in thumb-tab box for convenient handling

Manufacturer: General Data Healthcare™ SMSCTTW

 View all options for this product

Catalog No. 18-186-16


This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Color White
    Length (Metric) 75mm
    Quantity 72/Pk.
    Width (Metric) 25mm
    Coating Silane
    Thickness 1mm

    • Thumb-tab cutouts in box facilitate slide pickup
    • Insert slides directly into hoppers with one easy step
    • Contrasting dark color on bottom of slides enhances writing visibility
    • Available with or without Silane coating

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit