General Data Healthcare™ SHUR/Mark™ PLUS Color-Frosted Silane-Coated Inkjet Microscope Slides
Prepackaged in thumb-tab box for convenient pick-up and insertion into SHUR/Mark Slide Labeling Hoppers
Manufacturer: General Data Healthcare™ SMPSCTTB
 View more versions of this product
Catalog No. 22-044-684
  Check Availability
Add to Cart
    Specifications
    Color Blue
    Type Inkjet
    Adhesion Coating Silane
  • Description & Specifications
Related Products
Product Content Feedback