Learn More
General Data Healthcare™ SHUR/Mount™ Liquid Cover Glass and Mounting Medium
Convenient twist-top spout eliminates messy bottles and droppers
Manufacturer:  General Data Healthcare™ LC
 View more versions of this product
Catalog No. 15-184-40
Add to cart
Description
Description
  • Antioxidant prevents fading and UV absorbent prevents yellowing
  • Toluene-based acrylic resin allows rapid drying for hand coverslipping
  • Xylene-based for automated coverslipping instruments
  • Low viscosity minimizes bubble formation under cover glass
Specifications
Specifications
Liquid Cover Glass
4 oz.
118.2mL