Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ SHUR/Mount™ Liquid Cover Glass and Mounting Medium

Convenient twist-top spout eliminates messy bottles and droppers

$338.50 - $1015.00


Products 2
Catalog Number Mfr. No. Product Type Volume (English) Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Volume (English) Volume (Metric) Price Quantity & Availability  
15-184-40
View Documents
General Data Healthcare™
LC
Liquid Cover Glass 4 oz. 118.2mL
Case of 12 for $1,015.00
Add to cart
 
22-047-150
View Documents
General Data Healthcare™
LCA
Low-viscosity Mounting Medium 16 oz. 471mL
Case of 4 for $338.50
Add to cart
 
Description

Description

  • Antioxidant prevents fading and UV absorbent prevents yellowing
  • Toluene-based acrylic resin allows rapid drying for hand coverslipping
  • Xylene-based for automated coverslipping instruments
  • Low viscosity minimizes bubble formation under cover glass

SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit