Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

General Data Healthcare™ SHUR/Stain™ 3030 H&E Slide Stainer

Comes with continuous loading with multi-protocol management

Manufacturer:  General Data Healthcare™ SS3030120

Catalog No. 22-048-014


Add to cart

Description

Description

  • Compact footprint — only 34 in. (86cm) wide
  • 30 stations, five rinse stations, two water inlets
  • Number of programs: 50
  • State-of-the-art, user-friendly operator interface with color touch screen
  • Built-in fume extraction or can be connected to lab fume exhaust system
  • Display: Active matrix TFT 6.5 in. display
  • Voltage: AC Factory set for either 100-120 volts or 230 volts operation, 50 or 60 Hz
  • Fuse rating: 10 Amp x 250 volts Time Delay (Slo-Blo) fuse for 100-120 volts units

  • Number of Load Station: 1
  • Number of Unload Station: 2
  • Reagent Container Volume: 350mL
  • Slide Carrier Capacity: 30 Slides
  • Drying Station Temp Range: 40° to 70°C
  • Resistive touch pad
  • Programmable Flow Rates: 500, 1000, 1500, and 2000mL/min, programmable for individual step

Specifications

Specifications

Shur/Stain™ H And E Slide Stainer
30
40° to 70°C
65cm
57kg
50/60Hz
UL/FCC
100/240V 50/60Hz
350mL
26 in.
125 lb.
100/240V
30 stations, 5 rinse stations, 2 water inlets, interface with color touchscreen
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit