Learn More
General Data Healthcare™ SHUR/Track™ Process and Embedding Cassettes with Channel Track Hinge
Unique Track Hinge design provides easy one-handed installation of lid cover
$166.00
Specifications
Closure Type Separate Lid
Description Processing/Embedding Cassette
View More Specs
Products 6
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
22-048-037
View Documents
General Data Healthcare™
SMPPCG
Green
Case of 1000 for $166.00
Add to cart
22-048-038
View Documents
General Data Healthcare™
SMPPCGY
Gray
Case of 1000 for $166.00
Add to cart
22-048-054
View Documents
General Data Healthcare™
SMPPCW
White
Case of 1000 for $166.00
Add to cart
22-048-036
View Documents
General Data Healthcare™
SMPPCB
Blue This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
22-048-039
View Documents
General Data Healthcare™
SMPPCP
Pink This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
22-048-043
View Documents
General Data Healthcare™
SMPPCA
Aqua This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Front catch and four corner locking mechanism
  • Using rear track clasp, lid can be adjusted to stay open in place
  • Chemical resistant ridged materials provide a stable platform
  • For use with both automated labeling instruments and hand marking devices
  • Detachable lids are removed and packaged separately
Specifications
Specifications
Separate Lid
Processing/Embedding Cassette