General Data Healthcare™ TFM™ Colored Tissue Freezing Medium

Variety of colors to easily differentiate multiple specimens during sectioning and staining

Manufacturer: General Data Healthcare™ TFM5

 View more versions of this product

Catalog No. 15-183-36

  Check Availability
Add to Cart

    Specifications

    Boiling Point 212°C
    Special Use Embedding matrix for frozen sections
    Color Blue, clear, green, red, and yellow,Blue, clear, green, red, and yellow
    Product Type Tissue Freezing Medium™ 5 Color Kit
    Quantity 118mL,118mL
    Type Tissue Freezing Medium™ 5 Color Kit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit