Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBX3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Recombinant Polyclonal Antibody

Manufacturer:  LIFE TECHNOLOGIES 711087

Encompass

Catalog No. 71-108-7


Add to cart

Description

Description

This antibody is predicted to react with Monkey, Pig and Rat Recombinant rabbit polyclonal antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit polyclonal antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production.

TBX3 Recombinant Polyclonal antibody specifically detects TBX3 in Human samples. It is validated for ChIP assay, Immunocytochemistry, Immunofluorescence
Specifications

Specifications

TBX3
ChIP assay, Immunocytochemistry, Immunofluorescence
22HCLC
Unconjugated
Tbx3
bladder cancer related protein XHL; D5Ertd189e; T-box 3; T-box 3 (ulnar mammary syndrome); T-box protein 3; T-box transcription factor TBX3; Tbx3; TBX3-ISO; UMS; XHL
Protein corresponding to Human TBX3 (aa 305-500)
Protein A
RUO
Antibody
Polyclonal
Human
Liquid
Polyclonal
0.5 mg/mL
PBS with 0.09% sodium azide; pH 7.2
O15119
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
6926
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit