Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TBX3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | TBX3 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry |
Applications | Western Blot, Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB126841
|
Novus Biologicals
NBP310971100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
TBX3 Polyclonal specifically detects TBX3 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
TBX3 | |
Western Blot, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL | |
The immunogen is a synthetic peptide directed towards the middle region of human TBX3 (NP_057653). Peptide sequence VGTSNLKDLCPSEGESDAEAESKEEHGPEACDAAKISTTTSEEPCRDKGS | |
Affinity purified |
Western Blot 1.0 ug/ml, Immunohistochemistry | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
6926 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title