Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TBX3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP311386200UL

 View more versions of this product

Catalog No. NB121480


Add to cart

Description

Description

TBX3 Polyclonal antibody specifically detects TBX3 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

TBX3
Unconjugated
PBS, pH7.0
bladder cancer related protein XHL, T-box 3, T-box protein 3, T-box transcription factor TBX3, TBX3-ISO, ulnar mammary syndrome, UMS, XHL
E. coli-derived Human TBX3 fragment
Antigen and protein A Affinity-purified
RUO
Primary
6926
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Immunohistochemistry -1:50-1:200, Immunocytochemistry/Immunofluorescence -1:50-1:200, Immunohistochemistry-Paraffin -1:50-1:200
Purified
Rabbit
IgG
200 μg
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit