Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TBXA2R Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA534257
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Panda (100%) Gibbon, Monkey, Marmoset, Bovine (94%) Bat, Elephant (88%) Dog (81%).
The thromboxane A2 receptor (TBXA2R) is a member of the Prostanoid Receptor subfamily. TBXA2R performs an essential role in hemostasis by interacting with thromboxane A2 to induce platelet aggregation. Thromboxane A2 has been implicated in myocardial infarct, stroke, and asthma. Additionally, thromboxane-prostanoid receptors have been implicated in the pathogenesis of cardiovascular diseases. Changes in TBXA2R function have also been linked to a hereditary bleeding disorder.
Specifications
TBXA2R | |
Polyclonal | |
Unconjugated | |
TBXA2R | |
BDPLT13; prostanoid TP receptor; TBXA2R; Thromboxane A2 receptor; thromboxane prostanoid; Thromboxane receptor; TP; TXA2 receptor; TXA2-R; TXR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
6915 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
P21731 | |
TBXA2R | |
Synthetic 16 amino acid peptide from 1st cytoplasmic domain of human TBXA2R. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction