Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thromboxane synthase Mouse anti-Human, Clone: OTI1A9, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525163

Catalog No. PIMA525163


Add to cart

Description

Description

This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. However, this protein is considered a member of the cytochrome P450 superfamily on the basis of sequence similarity rather than functional similarity. This endoplasmic reticulum membrane protein catalyzes the conversion of prostglandin H2 to thromboxane A2, a potent vasoconstrictor and inducer of platelet aggregation. The enzyme plays a role in several pathophysiological processes including hemostasis, cardiovascular disease, and stroke. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications

Specifications

Thromboxane synthase
Monoclonal
0.86 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P24557
BDPLT14; CYP5; CYP5A1; Cytochrome P450 5A1; Cytochrome P450; family 5; subfamily A; polypeptide 1; FLJ52771; GHOSAL; Platelet cytochrome P450 subfamily V; TBXAS1; THAS; THAS_HUMAN; Thromboxane A synthase 1 (platelet); Thromboxane A synthase 1 (platelet; cytochrome P450; family 5; subfamily A); Thromboxane A synthase 1; Thromboxane A synthase 1 platelet cytochrome P450 subfamily V; Thromboxane-A synthase; TS; TXA synthase; TXAS; TXS;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
6916
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1A9
Unconjugated
TBXAS1
Liquid
TBXAS1
Full length human recombit protein of TBXAS1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit