Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC 1, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
High affinity σ1 receptor ligand that displays selectivity over σ2 receptors (Ki values are 10 and 370 nM respectively). Exhibits low affinity for dopamine (DAT), serotonin (SERT) and noradrenalin (NET) transporters (Ki > 10 μM) and low affinity for dopamine D2 receptors (Ki = 1226 nM). Attenuates cocaine-induced locomotor activity in mice.
Specifications
Quantity | 50 mg |
Formula Weight | 297.37 |
Percent Purity | >99% |
Chemical Name or Material | TC 1 |
Safety and Handling
Recommended Storage : Store at Room Temperature