Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TC 1698 dihydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
787587-06-8
C13H20Cl2N2
275.217
ZQUQBCHNSBJMCG-UHFFFAOYSA-N
tc 1698 dihydrochloride,2-3-pyridinyl-1-azabicyclo 3.2.2 nonane dihydrochloride,2-3-pyridyl-1-azabicyclo 3.2.2 nonane dihydrochloride
21094258
2-pyridin-3-yl-1-azabicyclo[3.2.2]nonane;dihydrochloride
C1CC(N2CCC1CC2)C3=CN=CC=C3.Cl.Cl
This item is not returnable. View return policy
787587-06-8
C13H20Cl2N2
275.217
ZQUQBCHNSBJMCG-UHFFFAOYSA-N
tc 1698 dihydrochloride,2-3-pyridinyl-1-azabicyclo 3.2.2 nonane dihydrochloride,2-3-pyridyl-1-azabicyclo 3.2.2 nonane dihydrochloride
21094258
2-pyridin-3-yl-1-azabicyclo[3.2.2]nonane;dihydrochloride
C1CC(N2CCC1CC2)C3=CN=CC=C3.Cl.Cl

α7-selective agonist

Selective nicotinic α7 receptor agonist (EC 50 = 440 nM). Also displays weak partial agonist/antagonist activity at β-subunit-containing receptors. Neuroprotective.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
Formula Weight 275.22
Percent Purity >98%
Chemical Name or Material TC 1698 dihydrochloride

Recommended Storage : Desiccate at Room Temperature