Learn More
Sklar InstrumentsSupplier Diversity Partner TC Arruga Needle Holder, Sklar, 6-1/4 in., Blunt, Straight, Cross-Serrated, Pliers with Spring, Tungsten Carbide / Stainless Steel. PK/12. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 213100
TC Arruga Needle Holder, Sklar, 6-1/4 in., Blunt, Straight, Cross-Serrated, Pliers with Spring, Tungsten Carbide / Stainless Steel. Sklar's TC Arruga Needle Holder is primarily used in ophthalmologic procedures such as eyelid surgery. It is a single spring instrument with a ratchet locking mechanism at the base of the rectangular plier handle. This product is straight with cross-serrated tungsten carbide tips. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. The overall length of this product is 6-1/4 inches.
Catalog No. 50-143-7596