Learn More
Sklar InstrumentsSupplier Diversity Partner TC Crile Wood Needle Holder, Sklar, 6 in., Blunt, Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 218003
TC Crile Wood Needle Holder, Sklar, 6 in., Blunt, Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's TC Crile-Wood Needle Holder is used to hold and guide small to medium size needles and suture materials. The Crile-Wood Needle Holder has a gently tapered, blunt tip and is generally more delicate than the Mayo-Hegar but slightly less delicate than Webster, Halsey, or DeBakey needle holders. The ratcheted finger ring handle aids in securing needles for both continuous and interrupted closures. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. This product is straight with smooth tips, and an overall length of 6 inches.
Catalog No. 50-143-7693