Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Edge TC Mayo Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 161515
Sklar Edge TC Mayo Dissecting Scissors, Sklar, 9 in., Blunt/Blunt, Straight, Smooth, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar Edge TC Mayo Dissecting Scissors are a modification of standard Mayo scissors that provide an exceptionally hard and durable cutting edge designed to stay sharp for a significantly longer period of time than standard scissors. Sklar permanently bonds the tungsten carbide inserts to the blade of the scissors which maximizes control and precision as well. Mayo Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade. While the curved instrument is most commonly utilized for cutting or dissecting dense or deep tissue, and the straight version is typically used for cutting surface tissue or sutures, both may be used interchangeably depending on procedure and physician preference.
Catalog No. 50-143-7380