Learn More
Tocris Bioscience™ TC-E 5001
Potent tankyrase inhibitor
Manufacturer:  Tocris Bioscience™ 5049/50
 View more versions of this product
Catalog No. 504950R
Add to cart
This item is not returnable. View return policy
Description
Description
Dual tankyrase (TNKS) inhibitor (Kd values are 79 and 28 nM for TNKS1 and TNKS2 respectively, IC50 = 33 nM for TNKS2) that is devoid of activity at PARP1 and PARP2 (IC50 >19 μM). Inhibits Wnt signaling and stabilizes Axin2 levels.