Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ TC-E 5001

Potent tankyrase inhibitor

$149.00 - $619.00

Specifications

Product Type TC-E 5001
Inhibitors TC-E 5001
Molecular Weight 409.46
Purity >98
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
504910R View Documents Tocris Bioscience™
5049/10
10mg Each for $149.00
Add to cart
 
504950R View Documents Tocris Bioscience™
5049/50
50mg Each for $619.00
Add to cart
 
Description

Description

Dual tankyrase (TNKS) inhibitor (Kd values are 79 and 28 nM for TNKS1 and TNKS2 respectively, IC50 = 33 nM for TNKS2) that is devoid of activity at PARP1 and PARP2 (IC50 >19 μM). Inhibits Wnt signaling and stabilizes Axin2 levels.
Specifications

Specifications

TC-E 5001
TC-E 5001
409.46
>98

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit