Tocris™ TC-E 5001

Potent tankyrase inhibitor

Manufacturer: Tocris™ 5049/10

 View more versions of this product

Catalog No. 504910R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors TC-E 5001
    Purity >98
    Molecular Weight 409.46
    Quantity 10mg
    Product Type TC-E 5001

    Dual tankyrase (TNKS) inhibitor (Kd values are 79 and 28nM for TNKS1 and TNKS2 respectively, IC50 = 33nM for TNKS2) that is devoid of activity at PARP1 and PARP2 (IC50 >19µM). Inhibits Wnt signaling and stabilizes Axin2 levels.

    • Chemical Name: 3-(4-Methoxyphenyl)-5-[[[4-(4-methoxyphenyl)-5-methyl-4H-1,2,4-triazol-3-yl]thio]methyl]-1,2,4-oxadiazole
    • Formula: C20H19N5O3S
    • CAS Number: 865565-29-3
    • Solubility: Soluble to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit