Tocris™ TC-E 5002

Selective KDM2/7 inhibitor

Manufacturer: Tocris™ 5089/10

 View more versions of this product

Catalog No. 50-891-0

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors TC-E 5002
    Purity >98
    Molecular Weight 285.38
    Quantity 10mg
    Product Type TC-E 5002

    Selective histone demethylase KDM2/7 subfamily inhibitor (IC50 values are 0.2, 1.2, 6.8, 55, 83, >100 and >120µM for KDM7A, KDM7B, KDM2A, KDM5A, KDM4C, KDM6A and KDM4A respectively). Inhibits growth of HeLa and KYSE-150 cancer cells in vitro.

    • Chemical Name: N-(9-Cyclopropyl-1-oxononyl)-N-hydroxy-β-alanine
    • Formula: C15H27NO4
    • CAS Number: 1453071-47-0
    • Solubility: Soluble to 100mM in 1eq. NaOH and to 100mM in DMSO
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit