Learn More
Sklar InstrumentsSupplier Diversity Partner TC Euphrate-Pasque Needle Holder, Sklar, 8 in., Blunt, Straight, Smooth, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 214963
TC Euphrate-Pasque Needle Holder, Sklar, 8 in., Blunt, Straight, Smooth, Tungsten Carbide / Stainless Steel. Sklar's Euphrate-Pasque TC Needle Holder is a ratcheted, finger ring instrument used for holding small to intermediate size sutures. Sklar permanently bonds tungsten carbide inserts to the jaws, which maximizes control and precision when maneuvering the instrument and the needle. Short jaws with longer shanks allow the user to suture in small surgical sites. This product has smooth jaws and an overall length of 8 inches.
Catalog No. 50-143-7655