Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC HSD 21, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent 17β-hydroxysteroid dehydrogenase type 3 (17β-HSD3) inhibitor (IC50 values are 6 and 40 nM at human and mouse 17β-HSD3 respectively). Displays no activity at 17β-HSD1, 17β-HSD2, ERα, androgen receptors or glucocorticoid receptors.
Specifications
Quantity | 50 mg |
Formula Weight | 422.32 |
Percent Purity | >99% |
Chemical Name or Material | TC HSD 21 |
Safety and Handling
Recommended Storage | Store at +4°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction