Learn More
Sklar InstrumentsSupplier Diversity Partner TC Par Needle Holder, Sklar, 4-1/2 in., Straight, Cross-Serrated, Delicate, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 217012
TC Par Needle Holder, Sklar, 4-1/2 in., Straight, Cross-Serrated, Delicate, Finger Rings / Dual Gold, Tungsten Carbide / Stainless Steel. Sklar's TC Par needle holder has a delicate pattern and serrated jaw to ensure a firm grip on the suture needle. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
Catalog No. 50-143-7672