Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TC-SP 14, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective S1P1 agonist (EC50 values are 0.042 and 3.47 μM for human S1P1 and S1P3 respectively). Attenuates immune response to antigen challenge and reduces circulating blood lymphocyte counts in a rodent model of delayed-type hypersensitivity. Orally bioavailable.
Specifications
Quantity | 10 mg |
Formula Weight | 450.5 |
Percent Purity | >98% |
Chemical Name or Material | TC-SP 14 |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction