Learn More
Sklar InstrumentsSupplier Diversity Partner TC Sternal Wire Twister, Sklar, 5 in., Blunt, Straight, Cross-Serrated, Tungsten Carbide / Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 219060
TC Sternal Wire Twister, Sklar, 5 in., Blunt, Straight, Cross-Serrated, Tungsten Carbide / Stainless Steel. Sklar's TC Sternal Wire Twister is used for twisting wire or stainless steel sutures together, most commonly when closing the sternum. The cross-serrated working surface firmly holds wire and the ratchet locks jaws into place so the wire can be twisted. TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments. Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision. TC instruments are identified by gold handles.
Catalog No. 50-143-7844