Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TC10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530573
Description
Recommended positive controls: HepG2. Predicted reactivity: Mouse (99%), Rat (99%), Zebrafish (94%), Xenopus Tropicalis (94%), Chicken (96%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
TC10 is a member of the RAS superfamily of small GTP-binding proteins (see HRAS, MIM 190020) involved in insulin-stimulated glucose uptake.
Specifications
| TC10 | |
| Polyclonal | |
| Unconjugated | |
| RHOQ | |
| ARHQ; epididymis secretory protein Li 42; HEL-S-42; ras homolog family member Q; ras homolog gene family, member Q; RASL7A; ras-like protein family member 7A; Ras-like protein TC10; RAS-like, family 7, member A; ras-related GTP-binding protein TC10; RHOQ; Rho-related GTP-binding protein RhoQ; small GTPase Tc10; Tc10; TC10A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23433 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| P17081 | |
| RHOQ | |
| Recombinant protein encompassing a sequence within the center region of human TC10. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction