Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher BioReagents™ TCA Deblock, 3% trichloroacetic acid (TCA) in methylene chloride (w/v), Fisher BioReagents
Specifications
For Use With (Application) | DNA synthesis |
---|---|
Formulation | TCA 3% in Methylene Chloride |
pH | <2 |
Product Type | Reagent |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
BP31504
|
4000 mL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Specifications
DNA synthesis | |
TCA 3% in Methylene Chloride | |
<2 | |
Reagent |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title