Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TCAP Monoclonal Antibody (OTI3E7), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA526993
Description
TCAP Monoclonal antibody specifically detects TCAP in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
TCAP | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O15273 | |
Tcap | |
Full length human recombinant protein of TCAP produced in E.coli | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E7 | |
Unconjugated | |
Tcap | |
19 kDa sarcomeric protein; AI503981; AJ223855; CMD1N; CMH25; LGMD2G; limb girdle muscular dystrophy 2G (autosomal recessive); Tcap; T-cap; TELE; telethonin; teneurin C-terminal associated peptide; titin cap protein; titin-cap; titin-cap (telethonin) | |
Mouse | |
Affinity Chromatography | |
RUO | |
8557 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction